Anti-DD2 Rabbit Polyclonal Antibody [clone: Q60]

Proveedor: Bioworld Technology
BS3804
BWRLBS3804EA 262 EUR
BWRLBS3804
Anti-DD2 Rabbit Polyclonal Antibody [clone: Q60]
Anticuerpos
Synthetic peptide, corresponding to amino acids 50-100 of Human DD2.

DD2 (Q60) pAb detects endogenous levels of DD2 protein.

Type: Primary
Antigen: DD2
Clonality: Polyclonal
Clone: Q60
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR