Anti-GRB2 Rabbit Polyclonal Antibody [clone: R178]

Proveedor: Bioworld Technology
BS3871
BWRLBS3871EA 262 EUR
BWRLBS3871
Anti-GRB2 Rabbit Polyclonal Antibody [clone: R178]
Anticuerpos
Synthetic peptide, corresponding to amino acids 150-199 of Human GRB2.

GRB2 (R178) pAb detects endogenous levels of GRB2 protein.

Type: Primary
Antigen: GRB2
Clonality: Polyclonal
Clone: R178
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR