Anti-NCAM2 Rabbit Polyclonal Antibody [clone: L296]

Proveedor: Bioworld Technology
BS3889
BWRLBS3889EA 262 EUR
BWRLBS3889
Anti-NCAM2 Rabbit Polyclonal Antibody [clone: L296]
Anticuerpos
Synthetic peptide, corresponding to amino acids 271-320 of Human NCAM2.

NCAM2 (L296) pAb detects endogenous levels of NCAM2 protein.

Type: Primary
Antigen: NCAM2
Clonality: Polyclonal
Clone: L296
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno NCAM2
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Clon L296
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR