Anti-p-RPS6 Rabbit Polyclonal Antibody [clone: S235]

Proveedor: Bioworld Technology
BS4745
BWRLBS4745EA 284 EUR
BWRLBS4745
Anti-p-RPS6 Rabbit Polyclonal Antibody [clone: S235]
Anticuerpos
Synthetic phosphopeptide derived from human Ribosomal Protein S6 around the phosphorylation site of Serine 235.

p-Ribosomal Protein S6 (S235) pAb detects endogenous levels of Ribosomal Protein S6 protein only when phosphorylated at Ser235.

Type: Primary
Antigen: p-RPS6
Clonality: Polyclonal
Clone: S235
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno p-RPS6
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Clon S235
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo
Western blot Yes
FluorescenciaInmunológica Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR