Anti-SRSF11 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS5927
BWRLBS5927EA 262 EUR
BWRLBS5927
Anti-SRSF11 Rabbit Polyclonal Antibody
Anticuerpos
Synthetic peptide, corresponding to amino acids 260-305 of Human SRSF11.

SRSF11 pAb detects endogenous levels of SRSF11 protein.

Type: Primary
Antigen: SRSF11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR