Anti-RPS27 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS60087
BWRLBS60087EA 262 EUR
BWRLBS60087
Anti-RPS27 Rabbit Polyclonal Antibody
Anticuerpos
A synthetic peptide corresponding to residues in Human RPS27.

RPS27 pAb detects endogenous levels of RPS27 protein.

Type: Primary
Antigen: RPS27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR