Anti-AXIN1 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS60118
BWRLBS60118EA 262 EUR
BWRLBS60118
Anti-AXIN1 Rabbit Polyclonal Antibody
Anticuerpos
A synthetic peptide corresponding to residues in Human AXIN1.

AXIN1 pAb detects endogenous levels of AXIN1 protein.

Type: Primary
Antigen: AXIN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno AXIN1
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR