Anti-Troponin T1 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS60260
BWRLBS60260EA 262 EUR
BWRLBS60260
Anti-Troponin T1 Rabbit Polyclonal Antibody
Anticuerpos
A synthetic peptide corresponding to residues in Human Troponin T1.

Troponin T1 pAb detects endogenous levels of Troponin T1 protein.

Type: Primary
Antigen: Troponin T1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR