Anti-VEGF-A Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS6496
BWRLBS6496EA 341 EUR
BWRLBS6496
Anti-VEGF-A Rabbit Polyclonal Antibody
Anticuerpos
Recombinant full length Human VEGF-A.

VEGFA pAb detects endogenous levels of VEGFA protein.

Type: Primary
Antigen: VEGF-A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR