Anti-Albumin Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS6520
BWRLBS6520EA 341 EUR
BWRLBS6520
Anti-Albumin Rabbit Polyclonal Antibody
Anticuerpos
Recombinant full length Human Albumin.

Albumin pAb detects endogenous levels of Albumin protein.

Type: Primary
Antigen: Albumin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR