Anti-PSMC3 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS7204
BWRLBS7204EA 341 EUR
BWRLBS7204
Anti-PSMC3 Rabbit Polyclonal Antibody
Anticuerpos
Recombinant full length Human PSMC3 .

PSMC3 pAb detects endogenous levels of PSMC3 protein.

Type: Primary
Antigen: PSMC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno PSMC3
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo
Western blot Yes
InmunoQuímica Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR