Anti-PSMC4 Rabbit Polyclonal Antibody

Proveedor: Bioworld Technology
BS7365
BWRLBS7365EA 341 EUR
BWRLBS7365
Anti-PSMC4 Rabbit Polyclonal Antibody
Anticuerpos
Recombinant full length Human PSMC4.

PSMC4 pAb detects endogenous levels of PSMC4 protein.

Type: Primary
Antigen: PSMC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR