Anti-HSP70 Mouse Monoclonal Antibody

Proveedor: Bioworld Technology
MB0105
BWRLMB0105EA 341 EUR
BWRLMB0105
Anti-HSP70 Mouse Monoclonal Antibody
Anticuerpos
Synthetic peptide, corresponding to the N-terminus of Human HSP70.

This antibody detects endogenous levels of Hsp70 and does not cross-react with related proteins

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: N-terminal
Host: Mouse
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno HSP70
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Reactividad Human
Huésped Mouse
Isotipo
Western blot Yes
Epítopo N-terminal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR