Cesio cloruro ≥98% Grade II, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: Cesium Chloride

C6914-1KG C6914-500G
C6914-1KGEA 483 EUR
C6914-1KG C6914-500G
Cesio cloruro ≥98% Grade II, Sigma-Aldrich®
Cesio cloruro

It is used for the preparation of electrically conducting glasses. Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.


Cesium chloride (CsCl) is used to treat various kinds of cancers. Oral and intravenous administration of CsCl is linked with seizures, cardiotoxicity, syncope and death. In rats, CsCl reactions displays several variations in blood chemistry, that are categorised by a reduction of total protein content, pH, an increase in the level of urea, creatinine, glucose and total hemoglobin. Solutions may contain slight haze.

Formula: CsCl
Pto. Ebullición: 1303 °C (1013 hPa)
Pto. de fusión: 645 °C (lit.)
Densidad: 3,988 g/cm³ (20 °C)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00010955
Núm. CAS: 7647-17-8
EINECS: 231-600-2
Índice Merck: 13,02021
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06c1f0c854f333da9fda7dbd589bcefde0fd75ac&vid=cfa19f1981c213c94a0aadf765affd3a5b5966b3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Colour) White
Appearance (Form) Powder
Solubility (Colour) Colorless
Solubility (Turbidity)
1 g/ml, H₂O
Clear
A280 UV Absorption
50% in H₂O
≤0.2
Sulfate (SO₄) ≤0.05 %
Titration with AgNO₃ ≥ 98 %
Recommended Retest Period 4 years

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR