Anti-Vtg Mouse Monoclonal Antibody [clone: CK-4B3]

Proveedor: Cayman Chemical
100012-100 100012-500
CAYM100012-100EA 546 EUR
CAYM100012-100 CAYM100012-500
Anti-Vtg Mouse Monoclonal Antibody [clone: CK-4B3]
Anticuerpos
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: CK-4B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: japanese medaka, killifish, Sheepshead Minnow,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR