Piceatannol

Proveedor: Cayman Chemical

10009366-10 10009366-25 10009366-5 10009366-50
CAYM10009366-10EA 130 EUR
CAYM10009366-10 CAYM10009366-25 CAYM10009366-5 CAYM10009366-50
Piceatannol
Piceatannol
Formula: C₁₄H₁₂O₄
Número MDL: MFCD00221715
Núm. CAS: 10083-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR