Anti-FABP3 Mouse Monoclonal Antibody [clone: CC68]

Proveedor: Cayman Chemical
10233-1
CAYM10233-1EA 460 EUR
CAYM10233-1
Anti-FABP3 Mouse Monoclonal Antibody [clone: CC68]
Anticuerpos
Type: Primary
Antigen: FABP3
Clonality: Monoclonal
Clone: CC68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR