2-Hydroxyestradiol

Proveedor: Cayman Chemical

13019-1 13019-5 13019-10 13019-25
CAYM13019-1EA 64.9 EUR
CAYM13019-1 CAYM13019-5 CAYM13019-10 CAYM13019-25
2-Hydroxyestradiol
2-Hydroxyestradiol
Formula: C₁₈H₂₄O₃
Número MDL: MFCD00010490
Núm. CAS: 362-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR