Anti-Methylated Lysine Rabbit Polyclonal Antibody (Biotin)

Proveedor: Cayman Chemical
13728-400
CAYM13728-400EA 866 EUR
CAYM13728-400
Anti-Methylated Lysine Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: Methylated Lysine
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: multi-species
Order Now

ESPECIFICACIONES

Simbolo antígeno Methylated Lysine
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Biotin
Reactividad
Huésped
Isotipo
Western blot Yes
ELISA Yes
Precipitación inmunológica Yes
Purification purified by affinity chromatography
Adsorción cruzada No
Storage buffer PBS, pH 7,0, containing 50% glycerol and 0,01% sodium azide
Temperatura de almacenamiento −20 °C
Shipping temperature −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR