Anti-NF kappaB (p65) Rabbit Polyclonal Antibody

Proveedor: Cayman Chemical
13751-1
CAYM13751-1EA 591 EUR
CAYM13751-1
Anti-NF kappaB (p65) Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NF kappaB (p65)
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Horse, Gorilla, Human, Monkey, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno NFKB3
Environmentally Preferable
Nombre del antígeno NF kappaB
Sinónimos de antígenos p65
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Horse,Monkey,Gorilla,Chimpanzee,Human,Bovine,Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
Inmunógeno a portion of the NF kappaB (p65) NLS
Purification purified by affinity chromatography
Adsorción cruzada No
Storage buffer PBS containing 0,05% BSA and 0,05% sodium azide
Temperatura de almacenamiento –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR