Cefoperazone sodium salt ≥95%, crystalline solid

Proveedor: Cayman Chemical
Peligro

16113-1 16113-5 16113-10
CAYM16113-1EA 71.5 EUR
CAYM16113-1 CAYM16113-5 CAYM16113-10
Cefoperazone sodium salt ≥95%, crystalline solid
Cefoperazone sodium salt

Cefoperazone is a cephalosporin antibiotic that is active in vitro against most Gram-positive and Gram-negative bacteria, including β-lactamase strains (3 – 4 µg/ml inhibits 90% of S. aureus growth). It has been studied in the treatment of upper and lower respiratory tract infections, urinary tract infections, gynecological infections, skin infections, as well as bacteremia.

Formula: C₂₅H₂₆N₉NaO₈S₂
Número MDL: MFCD07793331
Núm. CAS: 62893-20-3

Order Now

Resultados test especificaciones

Purity ≥95%
Formulation A crystalline solid
λmax 229, 268 nm
Storage –20 °C
Stability ≥2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR