(12Z)-9,10-Dihydroxy-12-octadecenoic acid ≥98.0%

Proveedor: Cayman Chemical

53400-25 53400-50 53400-100 53400-500
CAYM53400-25EA 47.3 EUR
CAYM53400-25 CAYM53400-50 CAYM53400-100 CAYM53400-500
(12Z)-9,10-Dihydroxy-12-octadecenoic acid ≥98.0%
(12Z)-9,10-Dihydroxy-12-octadecenoic acid

Leukotoxin is the 9(10) epoxide of linoleic acid, generated by neutrophils during the oxidative burst. This unstable compound is rapidly degraded by epoxide hydrolases to form the diol, (±)9(10)-DiHOME. Mitochondrial dysfunction, vasodilation and apoptosis are features of leukotoxin toxicity. In renal phydrolysesroximal tubular cells, the diol hydrolysis products of leukotoxin, such as (±)9(10)-DiHOME, have been directly implicated as the cytotoxic agent responsible for cell death.

Formula: C₁₈H₃₄O₄
Núm. CAS: 263399-34-4

Order Now

Resultados test especificaciones

Purity ≥98%
Formulation A solution in methyl acetate
Storage −20 °C
Stability 1 year from the QC date provided


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR