Pallets and sumps
Proveedor: CEMO
8529 7582 10138 7681 7683 10402 7685 10404 8530 7680 10139 7682 7684 10403 7686 10405 8531 7583 10140 7688 7690 10406 7692 10408 8532 7687 10141 7689 7691 10407 7693 10409 7397 7398 7399 7400
CEMG8529EA 352 EUR
CEMG8529 CEMG7582 CEMG10138 CEMG7681 CEMG7683 CEMG10402 CEMG7685 CEMG10404 CEMG8530 CEMG7680 CEMG10139 CEMG7682 CEMG7684 CEMG10403 CEMG7686 CEMG10405 CEMG8531 CEMG7583 CEMG10140 CEMG7688 CEMG7690 CEMG10406 CEMG7692 CEMG10408 CEMG8532 CEMG7687 CEMG10141 CEMG7689 CEMG7691 CEMG10407 CEMG7693 CEMG10409 CEMG7397 CEMG7398 CEMG7399 CEMG7400
Pallets and sumps
Almacén
The steel sumps provide safe storage for water-polluting as well as inflammable substances.
  • All SW sump pallets are equipped with hot-dip galvanised, removable gratings
  • A wide range of models ensures the correct solution for many application
  • Sump pallets with 100 mm ground clearance permit driving under from all sides
Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.