Thermal printer
Proveedor: CONSORT
AP414
CONSAP414EA 0 EUR
CONSAP414
Thermal printer
Impresoras Document Printers
This thermal printer can deliver both serial and parallel input when using RS232 and Centronix connectors respectively. Direct connection is available to connect all meters with digital output.
  • Prints 40 columns in normal and 80 columns in condensed condition
  • Roll length: ±28 m
Información de suministro: Supplied with manual, roll of thermal paper, mains adaptor (230 VAC) and RS232 cable.