Ion selective electrodes
Proveedor: CONSORT
ISE34B ISE23B ISE33B ISE35B ISE25B ISE24B ISE26B ISE27B
CONSISE34BEA 765 EUR
CONSISE34B CONSISE23B CONSISE33B CONSISE35B CONSISE25B CONSISE24B CONSISE26B CONSISE27B
Ion selective electrodes
Electrodos Electrodos selectivos de iones (ISE)
These combination electrodes have an epoxy body.
Solid state electrodes utilise relatively insoluble inorganic salts in a membrane. Solid state electrodes exist in homogeneous or heterogeneous forms. In both types, potentials are developed at the membrane surface due to the ion-exchange process.
Glass membrane electrodes are formed by the doping of the silicon dioxide glass matrix with various chemicals. The most common of the glass membrane electrodes is the pH electrode. Glass membrane electrodes are also available for the measurement of sodium ions.
Polymer membrane electrodesMore Product Information