Disinfectant spray, Contec® CyChlor
Proveedor: CONTEC
FBC500CC FBT100CC FBC100CC
CONTFBC500CCEA 128 EUR
CONTFBC500CC CONTFBT100CC CONTFBC100CC
Disinfectant spray, Contec® CyChlor
Soluciones de limpieza Desinfectantes
Stabilised Hypochlorous acid with purified water. Contec CyChlor is a new broad spectrum disinfectant designed for every day use.
  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination
  • Filtered to 0,2 μm and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Not