Cleanroom wipes, pre-saturated, PROSAT® Polywipe-C Heatseal
Proveedor: CONTEC
PSCS0009 PSCS0009-BPR PSCS0012-BPR
CONTPSCS0009EA 576 EUR
CONTPSCS0009 115-2786 115-2787
Cleanroom wipes, pre-saturated, PROSAT® Polywipe-C Heatseal
Toallitas Toallitas para salas limpias Presaturadas
The Polywipe-C Heatseal wipe is made from filament polyester and pre-saturated with 70% IPA and 30% DI water.
  • Exceptionally low in particles and extractable residue
  • Presaturation greatly reduces solvent usage and maximises process control and consistency
  • Abrasion and chemical resistant
  • Controlled saturation level greatly reduces solvent usage and waste
  • Reduces VOC's associated with IPA
  • Consistent saturation maximises process control and efficiency
The wipe is laser cut to bond the fibers at the edges of the wipe. This creates a very clean, non-More Product Information