Detergent, Contec® Sterile NeutraKlean
Proveedor: CONTEC
SBC503NK SBT103NK SBC103NK
141-1306EA 269 EUR
141-1306 141-1305 CONTSBC103NK
Detergent, Contec® Sterile NeutraKlean
Soluciones de limpieza Detergentes y Limpiadores de Superficie
Contec® Sterile NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms. Ready to use, irradiated neutral detergent with purified water.
  • Neutral pH makes it suitable for use in all areas of cleanroom as gentle on surfaces
  • Filtered to 0.2 microns under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates
  • Low-foaming detergent action is ideal for use on floors, walls, and benches
  • Low level of surfactant provides low residue which can be easily removed
  • Trigger