Magnetic stirring bars, oval
Proveedor: COWIE
001.650.17 001.670.27 001.650.RE 001.640.RE 001.635.RE 001.630.10 001.625.RE 001.625.10 001.620.RE 001.615.RE 001.610.RE 001.664.RE 001.640.13 001.6100 001.6150
442-0125EA 6.2 EUR
442-0125 442-0126 COWI001.650.RE COWI001.640.RE COWI001.635.RE 442-0123 COWI001.625.RE 442-0122 COWI001.620.RE COWI001.615.RE COWI001.610.RE COWI001.664.RE COWI001.640.13 COWI001.6100 COWI001.6150
Magnetic stirring bars, oval
Imánes de agitación Barras magnéticas de agitación
PTFE covered. For stirrers with rare earth magnetic core the magnetic attraction is 1,5 to 2 times stronger than conventional stirrers. The magnetic core is made from a combination of samarium and cobalt. The rare earth stirrers will continue to function at full strength for many years as the magnetism remains for longer.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)