Shaft guides, High Performance (HP), for standard taper ground glass joints
Proveedor: COWIE
005.00.0619 005.00.0624 005.00.0819 005.00.0824 005.00.1024 005.00.1029 005.00.1045 005.00.1229 005.00.1634 005.00.1645 005.00.1945 005.00.0824.PK 005.00.1024.PK 005.00.1029.PK 005.00.1045.PK 005.00.1229.PK 005.00.1634.PK 005.00.1645.PK 005.00.1945.PK 005.00.0819.PK PC710.0819.PK PC710.0825.PK PC710.1019.PK PC710.1025.PK PC710.1225.PK PC710.1625.PK PC710.1925.PK 005.0.1029 005.0.1945.PK 005.0.1945 005.0.1634.PK 005.0.1634 005.0.1229.PK 005.0.1229 005.0.1045.PK 005.0.1045 005.0.1034.PK 005.0.1029.PK 005.0.1024.PK 005.0.1024 005.0.0824.PK 005.0.0824 005.0.0819.PK 005.0.0819 005.0.0624 005.0.0619 005.0.1034
COWI005.00.0619EA 116 EUR
COWI005.00.0619 COWI005.00.0624 COWI005.00.0819 COWI005.00.0824 COWI005.00.1024 COWI005.00.1029 COWI005.00.1045 COWI005.00.1229 COWI005.00.1634 COWI005.00.1645 COWI005.00.1945 COWI005.00.0824.PK COWI005.00.1024.PK COWI005.00.1029.PK COWI005.00.1045.PK COWI005.00.1229.PK COWI005.00.1634.PK COWI005.00.1645.PK COWI005.00.1945.PK COWI005.00.0819.PK COWIPC710.0819.PK COWIPC710.0825.PK COWIPC710.1019.PK COWIPC710.1025.PK COWIPC710.1225.PK COWIPC710.1625.PK COWIPC710.1925.PK COWI005.0.1029 COWI005.0.1945.PK COWI005.0.1945 COWI005.0.1634.PK COWI005.0.1634 COWI005.0.1229.PK COWI005.0.1229 COWI005.0.1045.PK COWI005.0.1045 COWI005.0.1034.PK COWI005.0.1029.PK COWI005.0.1024.PK COWI005.0.1024 COWI005.0.0824.PK COWI005.0.0824 COWI005.0.0819.PK COWI005.0.0819 COWI005.0.0624 COWI005.0.0619 COWI005.0.1034
Shaft guides, High Performance (HP), for standard taper ground glass joints
Imánes de agitación Ejes de agitación
High Performance (HP) shaft guides provide an effective guide for use with glass and metal shaft stirrers over a wide range of temperatures without shedding particles from a specially formulated PTFE-PEEK seal whilst maintaining vacuum. The moulded-in PTFE-PEEK guide ring located at the end of the taper provides a guide to assist alignment of the shaft.
Shaft guides have an NPT thread for direct assembly into PTFE reactor lids or standard tapers for use with glass joints.
Precaución: PEEK has reduced chemical resistance compared to PTFE and may be affected by acids, phenols and halogenMore Product Information