Stoppers, self realasing, PTFE
Proveedor: COWIE
010.1219 010.1224 010.1229 010.1234 010.1245 010.1145 010.1134 010.1119 010.1124
COWI010.1219EA 18.2 EUR
COWI010.1219 COWI010.1224 COWI010.1229 COWI010.1234 COWI010.1245 COWI010.1145 COWI010.1134 COWI010.1119 COWI010.1124
Stoppers, self realasing, PTFE
Tapones Tapones de PE/LDPE/HDPE
Fit standard ground glass joints.
With sealing rings and self releasing ring.