Tubing connectors, 4-way
Proveedor: COWIE
016.1115.2 016.1114.9.2 016.1114.2 016.1113.7.2 016.1113.2 016.1112.5.2 016.1112.2
COWI016.1115.2EA 21.1 EUR
COWI016.1115.2 COWI016.1114.9.2 COWI016.1114.2 COWI016.1113.7.2 COWI016.1113.2 COWI016.1112.5.2 COWI016.1112.2
Tubing connectors, 4-way
Racores y conectores
4-way connectors, made of PTFE, bayonet type for tubings.
  • Very good chemical resistance
  • Very good temperature resistance