Taper fittings for standard glass joints
Proveedor: COWIE
PC704.2429.14 PC704.1926.08 PC704.1926.12 PC704.1926.125 PC704.1926.250 PC704.1926.312 PC704.1926.500 PC704.2429.06 PC704.2429.12 PC704.2429.16 PC704.2429.250 PC704.2429.312 PC704.2429.500 PC704.2429.560 PC704.2429.625 PC704.2932.12 PC704.2932.14 PC704.2429.08 PC704.1926.06 PC704.1926.03 PC704.2932.500 PC704.2932.16 PC704.4540.750 PC704.45401000 PC704.4540.560 PC704.4540.500 PC704.3435.560 PC704.2932.560 PC704.2932.625 PC704.2932.750 PC704.3435.12 PC704.3435.14 PC704.3435.16 PC704.3435.19 PC704.4540.625 PC704.3435.500 PC704.2932.19 PC704.3435.625 PC704.3435.750 PC704.34351000 PC704.4540.12 PC704.4540.14 PC704.4540.16 PC704.4540.19 PC704.4540.25 PC704.3435.25 PC704.1938.312 PC704.2440.625 PC704.2440.560 PC704.2440.500 PC704.2440.312 PC704.2440.250 PC704.2440.16 PC704.2440.14 PC704.2440.12 PC704.2440.08 PC704.2942.14 PC704.1938.500 PC704.2942.16 PC704.1938.250 PC704.1938.125 PC704.1938.12 PC704.1938.08 PC704.1938.06 PC704.1938.03 PC704.2440.06 PC704.3445.500 PC704.4550.625 PC704.4550.560 PC704.4550.500 PC704.4550.25 PC704.4550.19 PC704.4550.16 PC704.4550.14 PC704.4550.12 PC704.3445.750 PC704.2942.12 PC704.3445.560 PC704.3445.25 PC704.3445.19 PC704.3445.16 PC704.3445.14 PC704.3445.12 PC704.34451000 PC704.2942.750 PC704.2942.625 PC704.2942.560 PC704.2942.500 PC704.2942.19 PC704.3445.625 PC704.4550.750
COWIPC704.2429.14EA 42.9 EUR
COWIPC704.2429.14 COWIPC704.1926.08 COWIPC704.1926.12 COWIPC704.1926.125 COWIPC704.1926.250 COWIPC704.1926.312 COWIPC704.1926.500 COWIPC704.2429.06 COWIPC704.2429.12 COWIPC704.2429.16 COWIPC704.2429.250 COWIPC704.2429.312 COWIPC704.2429.500 COWIPC704.2429.560 COWIPC704.2429.625 COWIPC704.2932.12 COWIPC704.2932.14 COWIPC704.2429.08 COWIPC704.1926.06 COWIPC704.1926.03 COWIPC704.2932.500 COWIPC704.2932.16 COWIPC704.4540.750 COWIPC704.45401000 COWIPC704.4540.560 COWIPC704.4540.500 COWIPC704.3435.560 COWIPC704.2932.560 COWIPC704.2932.625 COWIPC704.2932.750 COWIPC704.3435.12 COWIPC704.3435.14 COWIPC704.3435.16 COWIPC704.3435.19 COWIPC704.4540.625 COWIPC704.3435.500 COWIPC704.2932.19 COWIPC704.3435.625 COWIPC704.3435.750 COWIPC704.34351000 COWIPC704.4540.12 COWIPC704.4540.14 COWIPC704.4540.16 COWIPC704.4540.19 COWIPC704.4540.25 COWIPC704.3435.25 COWIPC704.1938.312 COWIPC704.2440.625 COWIPC704.2440.560 COWIPC704.2440.500 COWIPC704.2440.312 COWIPC704.2440.250 COWIPC704.2440.16 COWIPC704.2440.14 COWIPC704.2440.12 COWIPC704.2440.08 COWIPC704.2942.14 COWIPC704.1938.500 COWIPC704.2942.16 COWIPC704.1938.250 COWIPC704.1938.125 COWIPC704.1938.12 COWIPC704.1938.08 COWIPC704.1938.06 COWIPC704.1938.03 COWIPC704.2440.06 COWIPC704.3445.500 COWIPC704.4550.625 COWIPC704.4550.560 COWIPC704.4550.500 COWIPC704.4550.25 COWIPC704.4550.19 COWIPC704.4550.16 COWIPC704.4550.14 COWIPC704.4550.12 COWIPC704.3445.750 COWIPC704.2942.12 COWIPC704.3445.560 COWIPC704.3445.25 COWIPC704.3445.19 COWIPC704.3445.16 COWIPC704.3445.14 COWIPC704.3445.12 COWIPC704.34451000 COWIPC704.2942.750 COWIPC704.2942.625 COWIPC704.2942.560 COWIPC704.2942.500 COWIPC704.2942.19 COWIPC704.3445.625 COWIPC704.4550.750
Taper fittings for standard glass joints
Racores y conectores
Compression fittings are manufactured from pure PTFE with a PEEK olive to give a secure grip.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
Compression fittings fit standard taper ground glass joints to hold items such as temperature probes and sampling tubes securely in place.