Anti-Actin (Ab-1) (JLA20) Mouse Monoclonal Antibody

Proveedor: Merck Millipore (Oncogene)
CP01-1
CP01-1EA 544 EUR
CP01-1
Anti-Actin (Ab-1) (JLA20) Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Actin (Ab-1) (JLA20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan
Order Now

ESPECIFICACIONES

Simbolo antígeno ACT1
Environmentally Preferable
Nombre del antígeno Actin
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Reactividad Multiple
Huésped Mouse
Isotipo
Western blot Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR