Anti-PSD-95 (Ab-2) (7E3-1B8) Mouse Monoclonal Antibody [clone: 7E3-1B8]

Proveedor: Merck Millipore (Oncogene)
CP35-100
CP35-100EA 623 EUR
CP35-100
Anti-PSD-95 (Ab-2) (7E3-1B8) Mouse Monoclonal Antibody [clone: 7E3-1B8]
Anticuerpos
Type: Primary
Antigen: PSD-95 (Ab-2) (7E3-1B8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno PSD-95
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 7E3-1B8
Reactividad Rat
Huésped Mouse
Isotipo
Western blot Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR