E.Z.N.A.® Plasmid DNA Mini Kit I, (V-spin)
Proveedor: OMEGA BIO-TEK
D6943-03
D6943-03EA 533 EUR
D6943-03
E.Z.N.A.® Plasmid DNA Mini Kit I, (V-spin)
Reactivos de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
Isolate up to 25 µg plasmid DNA from 1 to 5 ml bacterial cultures using mini spin columns.
  • Rapid - Purification of plasmid DNA in less than 30 minutes
  • Safe - No Phenol/chloroform extractions
  • Versatile - Spin and vacuum formats available
  • High-quality - DNA is suitable for a variety of downstream applications
The E.Z.N.A.® Plasmid DNA Mini Kit I is designed to isolate up to 25 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in less than 30 minutes. Plasmid DNA purification follows the alkaline-lysis method and is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.