Bed pan
Proveedor: DELTALAB
19586
DELT19586EA 178 EUR
DELT19586
Bed pan
Sistemas para diagnóstico Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Bed pan made of white PP, designed for disabled people, for the collection of urine and faeces.
  • Features rounded edges for more comfort.
  • L-shaped handle for a better handling
  • Allows a secure one-handed grip and avoids accidental spillage
  • Can be washed in automatic machines
  • Useful for hanging on handles and beds
Información de suministro: Supplied as individually wrapped and with instructions for use.