Safety Containers
Proveedor: DELTALAB
232821 232828 232826
DELT232821EA 5.9 EUR
DELT232821 DELT232828 DELT232826
Safety Containers
Recipientes Contenedores de objetos punzantes
Made from recycled plastic. Suitable for a specific purpose and adapted to the demands of all types of healthcare and technical departments like treatment rooms and labs and technical facilities. One-handed disconnection. Strong handle for transporting.
  • Puncture resistant
  • Autoclavable at 121 °C
Design that aims to reduce sharps injuries by bringing boxes as close as possible to the point of care in trays designed with multiple depressions. Sharps are disposed of safely at the 'point of use', eliminating many risks of injury.
Certificaciones: MeetsMore Product Information