Tubing, silicone, vacuum
Proveedor: DEUTSCH NEUMANN
3110713 3110412 3110612 3110309 3110818 3110715 3110515 3111020 3111222 3110717 3110816
228-1575EA 313 EUR
228-1575 228-1581 228-1582 DENE3110309 DENE3110818 DENE3110715 DENE3110515 DENE3111020 DENE3111222 DENE3110717 DENE3110816
Tubing, silicone, vacuum
Tuberías Tuberías de vacío/presión
Made of Silicone, translucent
  • Very good chemical resistance
  • Temperature range −60 to +200 °C
  • Flexible
Certificaciones: Meet standards BFR XV, FDA CFR 177,2600, Europ.Pharmacopoeia