Prisms
Proveedor: DIDALAB
POD010615
DIDAPOD010615EA 17.5 EUR
DIDAPOD010615
Prisms
Materiales educativos Materiales Educativos de Física Actividades de aprendizaje de óptica
Prism made of standard optical glass with index near to 1,52. Equilateral dispersing prisms are used for wavelength separation applications. Intended for demonstration experiments of light dispersion.