Disinfectant, Oxivir Plus spray
Proveedor: Diversey
100829235 100829234
DIVY100829235EA 0 EUR
DIVY100829235 DIVY100829234
Disinfectant, Oxivir Plus spray
Soluciones de limpieza Desinfectantes
Oxivir Plus spray is a powerful broad spectrum cleaner disinfectant. Ideal for contaminated body spillages, controlling pathogen outbreaks and cleaning high risk infection areas.
  • Safe on all acid resistant surfaces including upholstery and carpets
  • Woolsafe approved for soft furnishings
Certificaciones: EN 14476 Norovirus in 2 minutes. EN 1276 E. coli in 5 minutes. EN 1650 in 15 minutes.
Precaución: Use biocides safely. Always read the label and product information before use.