Soft Care Des E Alcohol Based Hand Disinfectant
Proveedor: Diversey
100962439
DIVY100962439EA 218 EUR
DIVY100962439
Soft Care Des E Alcohol Based Hand Disinfectant
Dispensadores de Jabón y Dispensadores de Jabón en Loción
Soft Care Des E is an alcohol based 71,5% hand disinfectant gel for waterless skin disinfection. Soft Care Des E is suitable for use in all areas, including healthcare and food processing environments.
  • Contains ethanol, a highly effective disinfectant
  • Suitable for surgical disinfection
  • Fragrance and colour-free; Suitable for use in the food processing industry
  • Contains an effective moisturiser preventing skin irritation and dehydration
Hand disinfectant based on ethanol. Ethanol is a disinfectant which is very effective against resident and transientMore Product Information
New Product