TASKI® Jontec Eternum High Gloss Floor Emulsion
Proveedor: Diversey
7512694
DIVY7512694EA 62.4 EUR
DIVY7512694
TASKI® Jontec Eternum High Gloss Floor Emulsion
Soluciones de limpieza Detergentes y Limpiadores de Superficie
High-gloss, slip resistant floor emulsion for most types of smooth and water resistant, hard floors. Its technology provides superior traffic resistance against scuffs and dirt. It ensures reduced maintenance costs to improve productivity. It will be ideal in all prestige and heavy traffic areas where ordinary emulsions require a high level of maintenance.
  • High-gloss, protective layer
  • Hard, durable and wear resistant protection
  • Excellent adhesion to most floor types
  • Slip resistant according to international standards (ASTM D-2047)
  • Easy to repair