Safety dispensing can
Proveedor: DUPERTHAL
3.108.09
DUEH3.108.09EA 278.05 EUR
DUEH3.108.09
Safety dispensing can
Contenedores
Safety can is used for convenient removal of flammable liquids.
  • Interior tin plated
  • Exterior red powder coated
  • Self closing openings
  • Automatic pressure compensation/vaccum compensation
  • High stability
  • Good media resistance
These safety can has a flame arrester in filling opening and tap and markings prepared according to GefstoffV and BGV A8.
Filling opening seal: Accopac, tap seal: Teflon