Alcohol metabolite standards, Ethyl-beta-D-glucuronides, CRM, Cerilliant®, Supelco®
Proveedor: Merck
E-016-1ML E-048-1ML
E-016-1MLEA 168 EUR
E-016-1ML E-048-1ML
Alcohol metabolite standards, Ethyl-beta-D-glucuronides, CRM, Cerilliant®, Supelco®
Normativas Estándares orgánicos
Ethyl ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alcohol consumption.
  • Snap-N-Spike®/Snap-N-Shoot®
These Snap-N-Spike® Reference solutions ares applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.
Precaución: Storage temperature: −20 °C (freezer).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25743683ac6e93acdcc385b8d044852c1a8ef007&vid=25743683ac6e93acdcc385b8d044852c1a8ef007&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27de808b9caf9dc2727c6cd5cb7fb4c8fd020456&vid=27de808b9caf9dc2727c6cd5cb7fb4c8fd020456&prid=web&clid=SAPDAM