Starter-Kit, SL 1011
Proveedor: EBRO
1340-5831 1340-6193 1250-1011
EBRO1340-5831EA 123 EUR
EBRO1340-5831 EBRO1340-6193 EBRO1250-1011
Starter-Kit, SL 1011
Registradores de datos
The SL1011 set by ebro, featuring the EBI 12-T101 data logger, PHX800 pH tester, and TDS3 conductivity meter, is designed for validating and monitoring bedpan washers in healthcare settings, ensuring hygiene standards.
  • Comprehensive validation set for bedpan washers
  • Includes EBI 12-T101 temperature data logger, PHX800 pH tester, and TDS3 conductivity meter
  • High-precision monitoring for temperature, pH, and conductivity
  • Ideal for healthcare hygiene checks
  • Compact and durable instruments with high IP protection
  • Traceable factory calibration certificate at –20