Robust Temperature Bottle Data Loggers, EBI 12-T26x
Proveedor: EBRO
1340-6618 1340-6619 1340-6620 1340-6621 1340-6622
EBRO1340-6618EA 2920 EUR
EBRO1340-6618 EBRO1340-6619 EBRO1340-6620 EBRO1340-6621 EBRO1340-6622
Robust Temperature Bottle Data Loggers, EBI 12-T26x
Registradores de datos
The data logger is specifically designed for monitoring pasteurization processes, particularly in beverage cans or bottles. The sensor needle is inserted externally into the beverage packaging and securely fastened.
  • Wide measuring range, –55 to +140 °C
  • High-quality PT1000 sensor (class A)
  • Rigid, external temperature sensor: Ø 6 mm, various lengths
  • Robust IP 68 protection class: waterproof and vapour-tight
  • M 10 thread on external sensor
The EBI 12-T26x data logger is designed for pasteurization control in beverage cans or bottles. Its Pt 1000 sensor,More Product Information