Organic reference standards, dorzolamides, Ph. Eur.

Proveedor: EDQM

Y0000750 Y0000727
EDQMY0000750EA 245 EUR
EDQMY0000750 EDQMY0000727
Organic reference standards, dorzolamides, Ph. Eur.
Normativas Estándares orgánicos

Order Now

ESPECIFICACIONES

Descripción Dorzolamide for system suitability, Ph. Eur. standard Dorzolamide impurity A, (4R,6R)-4-(ethylamino)-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulphonamide-7,7-dioxide, Ph. Eur. standard
Environmentally Preferable
CAS No. 130693-82-2 120279-95-0
Formula C₁₀H₁₇ClN₂O₄S₃ C₁₀H₁₆N₂O₄S₃

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR