Experimentation kits, mitochondrial DNA analysis using PCR
Proveedor: EDVOTEK
332
EDVO332EA 132 EUR
EDVO332
Experimentation kits, mitochondrial DNA analysis using PCR
Materiales educativos Materiales educativos de biología Actividades de aprendizaje de PCR
The mitochondria are thought to have evolved from a symbiotic relationship between prokaryotic and eukaryotic cells. Mitochondria have their own DNA and are only inherited via the maternal line. In this experiment, students amplify two regions of their mitochondrial DNA.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Información de suministro: Kit includes instructions, proteinase K, PCRMore Product Information