Sequencing the human microbiome
Proveedor: EDVOTEK
339-340
EDVO339-340EA 109 EUR
EDVO339-340
Sequencing the human microbiome
Materiales educativos Materiales educativos de biología Genetics Learning Activities
Learn the basics of DNA sequencing and sequence homology with this lab activity.
  • No sophisticated lab equipment needed
  • Provides enough material for 10 lab groups
  • Takes 45 minutes to complete
Humans live in a delicate balance with the microorganisms that live in and on their bodies. If this balance is disrupted, harmful bacteria can multiply and cause disease. In this experiment, students will read DNA sequence obtained from automated DNA sequencing technologies. The data will be analyzed using publicly available databases to identify the bacterialMore Product Information